Order Entry
Austria
ContactUsLinkComponent
Test tube holder, stainless steel
Test tube holder, stainless steel
Catalog # BOCH2282
Supplier:  BOCHEM
undefined
Test tube holder, stainless steel
Catalog # BOCH2282
Supplier:  BOCHEM

Specifications

  • Length:
    150 mm
  • Pk:
    1
  • Spanning area:
    10 - 25 mm

Specifications

About this item

This test tube holder is made from stainless steel (18/10). For tubes with a diameter from 10 to 25 mm.