F1-ClipTip™ Single-Channel Pipettes, Mechanical, Fixed/Variable Volume
The Thermo Scientific™ F1-ClipTip™ pipette features interlocking technology to ensure secure tip attachment. The pipettes work exclusively with Thermo Scientific ClipTip pipette tips, providing a unique tip interface that locks the tip in place, and ensures a complete seal with minimal tip application and ejection forces. Tips are locked firmly in place and will not fall off.
- Silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models
- Improved grip on the volume adjustment knob ensures effortless and precise adjustment
- Exclusive interlock design ensures each tip is clipped securely on the F1-ClipTip until it is released with a light touch
- Secure volume setting prevents undesirable volume drift during operation
- 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles
- Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of Repetitive Strain Injury (RSI)
- Super blow-out offers 150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50 µl models and below
- Improved small volume pipetting 0,1 to 2 µl model and improved versions 1 to 10 µl models allow optimised performance in your small volume applications
- Wide selection of ClipTip pipette tips and Compatible sizes with F1-ClipTip pipettes: 12,5 Ext, 20, 50, 200, 300, 300 Ext and 1000 µl
Thermo Scientific F1-ClipTip pipettes are warranted to be free from defects in material and workmanship for a product lifetime (10 years). This warranty is subject to conditions as stated in the attached warranty information document and require an online registration, may vary by country.
: Supplied with maintenance set (incl. grease and O-rings), tip fitting tool, ClipTip tip sample, shelf hanger, warning labels, calibration certificate, warranty certificate and user's guide (English, French, German, Spanish).
GLP kits are available as well.
GLP kits are available as well.
- Catalog No:
- 613-5519
- 613-5520
- 613-5528
- 613-5499
- 613-7448
- 613-5529
- 613-5518
- 613-5522
- 613-5523
- 613-5526
- 613-5497
- 613-5527
- 613-5498
- 613-5524
- 613-5525
- 613-6961
- 613-6960
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
GLP kits
Frequently Bought Together