Order Entry
Austria
ContactUsLinkComponent
Anti-BKV VP1 Mouse Monoclonal Antibody [clone: 3G2]
Anti-BKV VP1 Mouse Monoclonal Antibody [clone: 3G2]
  ABNOMAB3204-M23A
 :  Abnova
Anti-BKV VP1 Mouse Monoclonal Antibody [clone: 3G2]
  ABNOMAB3204-M23A
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen symbol:
    BKV VP1
  • Clonality:
    Monoclonal
  • Clone:
    3G2
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
  • Environmentally Preferable:
  • Epitope:
    Full-length
  • Gene ID:
    1489515
  • Sequence:
    MGTNSVDTAPTKRKGECPGAAPKKPKEPVQVPKLLIKGGVEVLEVKTGVDAITEVECFLNPEMGDPDENLRGFSLKLSAENGFSSDSPERKMLPCYSTARIPLPNLNEDLTCGNLLMWEAVTVQTEVIGITSMLNLHAGSQKVHEHGGGKPIQGSNFHFFAVGGDPLEMQGVLMNYRTKYPDGTITPKNPTAQSQVMNTDHKAYLDKNNAYPVECWVPDPSRNENTRYFGTFTGGENVPPVLHVTNTATTVLLDEQGVGPLCKADSLYVSAADICGLFTNSSGTQQWRGLARYFKIRLRKRSVKNPYPISFLLSDLINRRTQRVDGQPMYGMESQVEEVRVFDGTERLPGDPDMIRYIDKQGQLQTKML
  • Size:
    200 μL
  • Pk:
    200 µl

 

 

Mouse monoclonal antibody raised against a full-length recombinant BKV VP1.

Type: Primary
Antigen: BKV VP1
Clonality: Monoclonal
Clone: 3G2
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: