Anti-VZV gE Mouse Monoclonal Antibody [clone: VA030]
Catalog # ABNOMAB13662
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Varicella zoster virus gE
- Antigen symbol:VZV gE
- Clonality:Monoclonal
- Clone:VA030
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human herpesvirus (HHV)
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against varicella zoster virus (VZV) gE.
Type: Primary
Antigen: VZV gE
Clonality: Monoclonal
Clone: VA030
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human herpesvirus (HHV)