DNA clean up kits, GeneClean® Turbo Kits, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
GeneClean® Turbo Kit is used for purification of DNA fragments of sizes 0,1 kb to 300 kb from TAE or TBE buffered agarose gels, PCR reactions and other enzymatic solutions. Purified DNA can be used in enzymatic reactions, conventional or automated sequencing, cloning and microarray analysis.
GeneClean® technology simplifies the process of purifying DNA into three easy steps: Bind, Wash, and Elute. Ethanol precipitation is never required, and purified DNA is immediately ready for a wide variety of downstream applications.
Delivery information: Kit contains GeneClean® Turbo Salt Solution, GeneClean® Turbo Wash Concentrate, GeneClean® Turbo Cartridges, GeneClean® Turbo Catch Tubes, GeneClean® Turbo Elution Solution, and GeneClean® Turbo GNomic Salt Solution.
Caution: For research purposes only.
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:DNA gel bandsAmplified/modified DNA
- Sample type:Gel matrixPCR and other enzymatic reactions
- Target molecule:DNA
- Description:GeneClean® Turbo Kit
Specifications
Frequently Bought Together