Order Entry
Austria
ContactUsLinkComponent
Fast RNA™ Pro Soil-Direct Kit, MP Biomedicals
Fast RNA™ Pro Soil-Direct Kit, MP Biomedicals
Catalog # ICNA116070050
Supplier:  MP Biomedicals
CAS Number:  
Fast RNA™ Pro Soil-Direct Kit, MP Biomedicals
Catalog # ICNA116070050
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Soil
  • Target molecule:
    RNA
  • Description:
    Fast RNA™ Pro Soil-Direct Kit, 50×2 ml preps
  • Description:
    Fast RNA™ Pro Soil-Direct Kit

Specifications

About this item

FastRNA™ Pro Soil-Direct Kit is used to isolate total RNA from organic material found in soil samples. The total RNA isolated is suitable for RT-PCR analysis and other downstream applications.

For research purposes only.

Delivery: Kit contains RNApro™ Soil Lysis Solution, 55 ml; Phenol:Chloroform (1:1), 50 ml; Inhibitor Removal Solution, 12 ml; RNAMATRIX® Slurry, 0,6 ml; RNAMATRIX® Binding Solution, 35 ml; RNAMATRIX® Wash Solution Concentrate, 15 ml; DEPC-treated H2O, 30 ml; Lysing Matrix E, 50×2 ml tubes; and Quick-Clean Spin Filters.