Order Entry
Austria
ContactUsLinkComponent
Fast RNA™ Pro green kit, MP Biomedicals
Fast RNA™ Pro green kit, MP Biomedicals
  ICNA116045050
 :  
undefined
Fast RNA™ Pro green kit, MP Biomedicals
  ICNA116045050
 :  

 

  • Pk:
    1 KIT
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Plant samples
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Description:
    Fast RNA™ Pro Green Kit, 50×2 ml preps
  • Description:
    Fast RNA™ Pro Green Kit

 

 

FastRNA® Pro Green Kit is used to isolate total RNA from any type of plant and animal tissue or cultured cells. Extracted RNA is suitable for RT-PCR analysis and other downstream applications.

The FastRNA™ Pro Green Kit is designed to efficiently isolate total RNA from any type of plant and animal tissue or cultured cells. Using the FastPrep-24™ or FastPrep® FP120 instrument, between 50 to 500 mg of tissue can be homogenised by Lysing Matrix D in impact-resistant 2 ml tubes. Total RNA is released into the proprietary, protective RNApro™ Solution. Following extraction with chloroform and precipitation, total RNA is extracted.

For research purposes only.

 : Kit contains RNApro™ Solution, 1×55 ml; DEPC-H2O, 1×15 ml; and Lysing Matrix D, 50×2 ml tubes.