Order Entry
Austria
ContactUsLinkComponent
Anti-PTPRC Rat Monoclonal Antibody (APC-Cyanine7) [clone: RA3-6B2]
Anti-PTPRC Rat Monoclonal Antibody (APC-Cyanine7) [clone: RA3-6B2]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-PTPRC Rat Monoclonal Antibody (APC-Cyanine7) [clone: RA3-6B2]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 25-0452-U100
  • 25-0452-U025
  • Antigen symbol:
  • CD45R
  • CD45R
  • Antigen name:
  • CD45R, or B220, a protein tyrosine phosphatase
  • CD45R, or B220, a protein tyrosine phosphatase
  • Antigen synonyms:
  • Lyt-4|Ly-5|T200
  • Lyt-4|Ly-5|T200
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)-Cy7®
  • APC (Allophycocyanin)-Cy7®
  • Clone:
  • RA3-6B2
  • RA3-6B2
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 19264
  • 19264
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • +2 °C to +8 °C, protected from light
  • +2 °C to +8 °C, protected from light

Specifications

Related Information

Frequently Bought Together