Order Entry
Austria
ContactUsLinkComponent
 
Anti-CD3 Mouse Monoclonal Antibody (APC-Cyanine7) [clone: SK7]
 
undefined
Anti-CD3 Mouse Monoclonal Antibody (APC-Cyanine7) [clone: SK7]

 

  • Catalog No:
  • 25-0036-T025
  • 25-0036-T100
  • Antigen symbol:
  • CD3
  • CD3
  • Antigen name:
  • CD3 epsilon subunitT cell receptor complex
  • CD3 epsilon subunitT cell receptor complex
  • Antigen synonyms:
  • Leu-4|T3
  • Leu-4|T3
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)-Cy7®
  • APC (Allophycocyanin)-Cy7®
  • Clone:
  • SK7
  • SK7
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 916
  • 916
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • +2 °C to +8 °C, protected from light
  • +2 °C to +8 °C, protected from light

 

Frequently Bought Together