Order Entry
Austria
Orders LinkContactUsLinkComponent
Group Lockout Boxes

Group Lockout Boxes

Supplier:   Brady
Group Lockout Boxes
Supplier:  Brady
Certificates
Group Lockout Boxes
Group Lockout Boxes

3 Options Available Below

Product Details & Documents

For group lockout situations involving a large number of workers and equipment. Can also be used as a lock storage box.

  • Transparent front cover allows for easy viewing of contents
  • Hooks for keys and slot for locks
  • Components not included
Product Family Options

Product Information

  • Colour
    W×D×H
    Material
    Availability
    Price
  • BRDY800127
    Group lockout centre with 18 hooks and 2 slots
    Group lockout centre with 18 hooks and 2 slots
    Red
    406×64×195 mm
    Acrylic
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 235,00 €
    235,00 €
    1 * 1 ST
       235,00 €

  • BRDY800116
    Group lockout centre with 8 hooks and 1 slot
    Group lockout centre with 8 hooks and 1 slot
    Red
    305×64×195 mm
    Acrylic
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 222,00 €
    222,00 €
    1 * 1 ST
       222,00 €

  • BRDY800117
    Group lockout centre with 3 hooks and 1 slot
    Group lockout centre with 3 hooks and 1 slot
    Red
    150×80×150 mm
    Acrylic
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 200,00 €
    200,00 €
    1 * 1 ST
       200,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.