ContactUsLinkComponent
Signs, floor signs, ToughStripe™ , dots
Signs, floor signs, ToughStripe™ , dots
Catalog # BRDY104400
Supplier:  Brady
undefined
Signs, floor signs, ToughStripe™ , dots
Catalog # BRDY104400
Supplier:  Brady
Specifications
  • Description:
    Floor marking dots, toughstripe floor making polyester, red
  • Dimensions:
    89 mm
  • Material:
    Polyester
  • Environmentally Preferable:
  • Pk:
    1 Roll
Specifications
About this item
ToughStripe™ products are used for marking aisles, passageways and storage locations, and pointing out important safety equipment and egress paths.
  • Floor marking products are made with B-514 polyester
  • Recommended for indoor use and dry environment
  • Resistant to harsh chemicals