Order Entry
Austria
ContactUsLinkComponent
Initial calibration standards
Initial calibration standards
Catalog # PERCN9303825
Supplier:  PerkinElmer
CAS Number:  
Initial calibration standards
Catalog # PERCN9303825
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pk:
    1
  • Description:
    Initial calibration verification standard 1, (5% HNO3/tr Tartaric Acid), 125 ml, 1000 mg/l: Ca, Fe, K, Mg, Na, Sr and 10 mg/l: Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Se, Tl, V, Zn, Th, U
  • Seq:
    01

Specifications

About this item