Orders LinkContactUsLinkComponent
Buckets, Rectangular
Buckets, Rectangular
Buckets, Rectangular
Certificates
Buckets, Rectangular
3 Options Available Below
Product Details & Documents
Standard bucket, rectangular, made of PP, with stainless steel handle, for Clino® system trolleys.
  • With scaling 5 to15 L
  • Coloured to facilitate use as part of a colour coded system
  • Good chemical resistance
  • Good temperature resistance
  • Autoclavable (approximately 15 to 30 cycles)
Documents
Product Family Options
Product Information
  • Colour
    Capacity
    Availability
    Price
  • Anthracite
    15 L
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 68,41 €
    68,41 €
    1 * 1 ST
       68,41 €
  • Blue
    15 L
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 68,41 €
    68,41 €
    1 * 1 ST
       68,41 €
  • Red
    15 L
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 68,41 €
    68,41 €
    1 * 1 ST
       68,41 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Cover for 15 l buckets, transparent
    Cover for 15 l buckets, transparent
    PPSA2000155
    Log in for availability
    1 * 1 ST - 44,91 €