ContactUsLinkComponent
Cleanroom mops, standard, MicroMopp CR/A
Cleanroom mops, standard, MicroMopp CR/A
Catalog # PPSA3500067
undefined
Cleanroom mops, standard, MicroMopp CR/A
Catalog # PPSA3500067
Specifications
  • Description:
    MicroMopp Standard CR/A mops, 110 g
  • Length:
    400 mm
  • Pk:
    1
Specifications
About this item
MicroMopp Standard CR/A mops are special multiple-use cleanroom mops.
  • Completely closed processing of polyester microsnares
  • 100 % PES-microfiber with polypropylene bristle
  • Rectangular mop design to reach edges perfectly well
  • Very good slide characteristics, not suitable for chlorinated disinfectants
  • For dissolving adhering contaminations and suitable for impregnation with flat wringer
  • With water-permeable cover and PU backfilling four enormous sqm-performance
  • Polyester pocket with drainage function