Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Environmental Test Kits
Powder pillows, cyanuric acid
Powder pillows, cyanuric acid
Powder pillows, cyanuric acid
Catalog # 705-0143
Supplier:
Hach
CAS Number:
Expand image
Powder pillows, cyanuric acid
Catalog # 705-0143
Supplier:
Hach
CAS Number:
Specifications
Chemical method:
Turbidimetric
Pk:
50 Tests
Description:
Cyanuric acid
Type:
PP
Range:
5 - 50 mg/l
Specifications
About this item
Product Details
Documentation
Product Details
Documentation