Capillary columns, MXT®-65TG
Supplier: Restek
Certificates
About this item
MXT®-65TG columns are application-specific columns, specially tested for triglycerides.
- High-polarity phase; Crossbond® diphenyl dimethyl polysiloxane
- Stable to 370 °C
- Coil diameter: 114 mm
The MXT®-65TG phase resolves triglycerides by degree of unsaturation as well as by carbon number. Because of the chemistry required to achieve 370 °C thermal stability, an MXT®-65TG column should not be used for analyses of compounds that contain active oxygenated groups.
Frequently Bought Together