ContactUsLinkComponent
Test kits, sodium hypochlorite, CHECKIT®
Catalog # LOVI147490
Supplier:  Lovibond
CAS Number:  
undefined
Test kits, sodium hypochlorite, CHECKIT®
Catalog # LOVI147490
Supplier:  Lovibond
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Sodium hypochlorite test kit
  • Range:
    2 - 18%
Specifications
About this item
CHECKIT® Comparator is a compact and handy colorimetric unit which is suitable for both mobile and static analysis work.
  • Easy operation and exact reagent dosing
  • Tablet reagents have minimum guaranteed shelf life of 5/10 years
The operating instructions provide a step-by-step explanation of conducting the water test to achieve reliable and accurate measurements in the minimum of time.
Delivery: Test kit includes CHECKIT® discs, cells, stirring rod and AQUALYTIC® tablet reagents (for 30 tests) for the desired test supplied in a sturdy plastic case.