Orders LinkContactUsLinkComponent
Test kits, chlorine (DPD), CHECKIT®
Supplier:   Lovibond
Test kits, chlorine (DPD), CHECKIT®
Supplier:  Lovibond
Certificates
Test kits, chlorine (DPD), CHECKIT®
Test kits, chlorine (DPD), CHECKIT®
13 Options Available Below
Product Details & Documents
CHECKIT® Comparator is a compact and handy colorimetric unit which is suitable for both mobile and static analysis work. Supplied with a generous number of different colour scales, it provides the basis for a comprehensive, easy-to use colorimetric analysis system.
  • Enables the use of large path lengths
  • The mirror optics makes use of the view through the entire length of the cell
  • Easy operation and exact reagent dosing
  • Tablet reagents have minimum guaranteed shelf life of 5/10 years
The operating instructions provide a step-by-step explanation of conducting the water test to achieve reliable and accurate measurements in the minimum of time.
CHECKIT® disc contains a continuous colour scale which makes it possible to achieve an exact colour match between the colour standard and the sample. These discs are specially manufactured in selected materials to remain colour-stability over a long period and guarantee reliable, reproducible measurement results.
 : Test kit includes CHECKIT® discs, cells, stirring rod and AQUALYTIC® tablet reagents (for 30 tests) for the desired test supplied in a sturdy plastic case.
Product Family Options
Product Information
  • Range
    Pk
    Availability
    Price
  • LOVI147000
    Chlorine test kit, DPD, (free, combined, total)
    Chlorine test kit, DPD, (free, combined, total)
    0,02 - 0,3 mg/l Cl2
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 76,20 €
    76,20 €
    1 * 1 KIT
       76,20 €
  • LOVI147010
    Chlorine test kit, DPD, (free, combined, total)
    Chlorine test kit, DPD, (free, combined, total)
    0 - 1 mg/l Cl2
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 67,50 €
    67,50 €
    1 * 1 KIT
       67,50 €
  • LOVI147040
    Chlorine test kit, DPD, (free, combined, total)
    Chlorine test kit, DPD, (free, combined, total)
    0 - 2 mg/l Cl2
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 67,50 €
    67,50 €
    1 * 1 KIT
       67,50 €
  • LOVI147020
    Chlorine test kit, DPD, (free, combined, total)
    Chlorine test kit, DPD, (free, combined, total)
    0 - 4 mg/l Cl2
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 67,50 €
    67,50 €
    1 * 1 KIT
       67,50 €
  • LOVI147052
    Chlorine test kit, DPD (free + total), powder pack
    Chlorine test kit, DPD (free + total), powder pack
    0 - 3,5 mg/l Cl2
    50 Tests
    Log in for availability
    Log in for availability
    1 * 50 Tests
    1 * 50 Tests - 96,30 €
    96,30 €
    1 * 50 Tests
       96,30 €
  • LOVI147051
    Chlorine test kit, DPD, (total), powder pack
    Chlorine test kit, DPD, (total), powder pack
    0 - 3,5 mg/l Cl2
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 96,30 €
    96,30 €
    1 * 1 KIT
       96,30 €
  • LOVI147050
    Chlorine test kit, DPD (free), powder pack
    Chlorine test kit, DPD (free), powder pack
    0 - 3,5 mg/l Cl2
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 96,30 €
    96,30 €
    1 * 1 KIT
       96,30 €
  • LOVI146000
    Chlorine disc, DPD (free, combined, total)
    Chlorine disc, DPD (free, combined, total)
    0,02 - 0,3 mg/l Cl2
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 27,70 €
    27,70 €
    1 * 1 KIT
       27,70 €
  • LOVI146010
    Chlorine disc, DPD (free, combined, total)
    Chlorine disc, DPD (free, combined, total)
    0 - 1 mg/l Cl2
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 27,70 €
    27,70 €
    1 * 1 KIT
       27,70 €
  • LOVI146040
    Chlorine disc, DPD (free, combined, total)
    Chlorine disc, DPD (free, combined, total)
    0 - 2 mg/l Cl2
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 27,70 €
    27,70 €
    1 * 1 KIT
       27,70 €
  • LOVI146050
    Chlorine disc, DPD (free, combined, total)
    Chlorine disc, DPD (free, combined, total)
    0 - 3,5 mg/l Cl2
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 27,70 €
    27,70 €
    1 * 1 KIT
       27,70 €
  • LOVI146020
    Chlorine disc, DPD (free, combined, total)
    Chlorine disc, DPD (free, combined, total)
    0 - 4 mg/l Cl2
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 27,70 €
    27,70 €
    1 * 1 KIT
       27,70 €
  • LOVI146030
    Chlorine disc, KI (total)
    Chlorine disc, KI (total)
    10 - 300 mg/l Cl2
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 27,70 €
    27,70 €
    1 * 1 KIT
       27,70 €
Accessories
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • CHECKIT® Comparator, path length 13,5 mm
    CHECKIT® Comparator, path length 13,5 mm
    LOVI145000
    1
    Log in for availability
    1 * 1 ST - 18,20 €
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • CHECKIT® plastic cells, pack of 10, with lid, 10 ml, 13,5 mm path length
    CHECKIT® plastic cells, pack of 10, with lid, 10 ml, 13,5 mm path length
    LOVI145500
    10
    Log in for availability
    1 * 10 ST - 41,30 €
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • CHECKIT® plastic cells, pack of 5, with lid, 10 ml, 13,5 mm path length
    CHECKIT® plastic cells, pack of 5, with lid, 10 ml, 13,5 mm path length
    LOVI145505
    5
    Log in for availability
    1 * 5 ST - 24,40 €
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • CHECKIT® plastic cells, pack of 100, with lid, 10 ml, 13,5 mm path length
    CHECKIT® plastic cells, pack of 100, with lid, 10 ml, 13,5 mm path length
    LOVI145510
    100
    Log in for availability
    1 * 100 ST - 342,00 €