Order Entry
Austria
Orders LinkContactUsLinkComponent
Electrode cleaning solution, RENOVO.N

Electrode cleaning solution, RENOVO.N

Mild cleaning solution for electrodes, RENOVO.N, 250 ml bottle, 250 mL
Catalog # HACHS16M001
Supplier:   Hach
Electrode cleaning solution, RENOVO.N
Catalog # HACHS16M001
Supplier:  Hach
CAS Number:  
Electrode cleaning solution, RENOVO.N
Electrode cleaning solution, RENOVO.N

Mild cleaning solution for electrodes, RENOVO.N, 250 ml bottle, 250 mL

CAS Number:  
Price
26,50 €
Log in for availability
1 * 250 mL
Product Details & Documents

Mild cleaning solution for electrodes. Ideal for use after measurements in aqueous solutions.

 

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Mild cleaning solution for electrodes, RENOVO.N, 250 ml bottle

Specifications