Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Electrode cleaning solution for proteins/organic samples

Electrode cleaning solution for proteins/organic samples, 250 ml bottle, 250 mL
Catalog # HACHC20C370
Supplier:   Hach
Electrode cleaning solution for proteins/organic samples
Catalog # HACHC20C370
Supplier:  Hach
CAS Number:  
Electrode cleaning solution for proteins/organic samples
Electrode cleaning solution for proteins/organic samples

Electrode cleaning solution for proteins/organic samples, 250 ml bottle, 250 mL

CAS Number:  
Price
44,70 €
Log in for availability
1 * 250 mL
Product Details & Documents

Electrode cleaning solution to remove proteins residues organic contaminations/deposits such as food stuff from pH sensing element.

Formula: PEPSIN in HCl

 

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Electrode cleaning solution for proteins/organic samples, 250 ml bottle

Specifications