Order Entry
Austria
Orders LinkContactUsLinkComponent
Storage Boxes for Tubes

Storage Boxes for Tubes

Supplier:   NEOLAB
Storage Boxes for Tubes
Supplier:  NEOLAB
Certificates
Storage Boxes for Tubes
Storage Boxes for Tubes

2 Options Available Below

Product Details & Documents

Made of PP, coloured.

  • Tight-closing hinged lid
  • Autoclavable at 121 °C
  • Good chemical resistance

Stackable storage boxes with 5×10 compartments accommodate vials up to 12 mm Ø with a max. height of 46 mm, such as HPLC vials, cryogenic vials, 1,5 and 2,0 ml microcentrifuge tubes.

Product Family Options

Product Information

  • Compartments
    Colour
    W×D×H ext.
    Availability
    Price
  • 5×10
    Yellow
    140×86×55 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,64 €
    11,64 €
    1 * 1 ST
       11,64 €

  • 5×10
    Pink
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,64 €
    11,64 €
    1 * 1 ST
       11,64 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.