ContactUsLinkComponent
Turbine spinner for use with Bruker® Spectrometers, SP Wilmad-LabGlass
Turbine spinner for use with Bruker® Spectrometers, SP Wilmad-LabGlass
Catalog # WILMB-PEEK-5
Supplier:  SP Industries
undefined
Turbine spinner for use with Bruker® Spectrometers, SP Wilmad-LabGlass
Catalog # WILMB-PEEK-5
Supplier:  SP Industries
Specifications
  • Description:
    Bruker spinner turbine, PEEK with Viton™ O-rings, 5 mm, suitable for application temperature range: -150... +200 °C
  • Material:
    PEEK
  • Pk:
    1
Specifications
About this item
Manufactured to precision standards that exceed the spectrometer manufacturers requirements.
  • Variable temperature operation of -150 to +200 °C
  • Weight compatible with room temperature spinners
  • Susceptibility matched with Bruker® originals
Can be mixed with other turbines during sample changer operation.