ContactUsLinkComponent
General purpose gloves, Senti-Flex, A360
General purpose gloves, Senti-Flex, A360
Supplier:  Portwest
Certificates
undefined
General purpose gloves, Senti-Flex, A360
Supplier:  Portwest
Specifications
  • Catalog No:
  • 112-4595
  • 112-4598
  • 112-4596
  • 112-4597
  • 112-4594
  • Model:
  • Senti-Flex, A360
  • Senti-Flex, A360
  • Senti-Flex, A360
  • Senti-Flex, A360
  • Senti-Flex, A360
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Nylon
  • Nylon
  • Nylon
  • Nylon
  • Nylon
  • Finish:
  • PU coating on palm and fingertips
  • PU coating on palm and fingertips
  • PU coating on palm and fingertips
  • PU coating on palm and fingertips
  • PU coating on palm and fingertips
  • Gauge:
  • 18
  • 18
  • 18
  • 18
  • 18
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (3121X)
  • EN 388:2016 (3121X)
  • EN 388:2016 (3121X)
  • EN 388:2016 (3121X)
  • EN 388:2016 (3121X)