Order Entry
Austria
ContactUsLinkComponent
Capillary collection tubes
Capillary collection tubes
Catalog # DRUM1-000-800/12
Supplier:  DRUMMOND
Capillary collection tubes
Catalog # DRUM1-000-800/12
Supplier:  DRUMMOND

Specifications

  • Ø int.:
    0,4 mm
  • Description:
    Capillary Tubes
  • Length:
    75 mm
  • Pk:
    100

Specifications

About this item

For coagulation, hemoglobin, prothrombin, blood grouping and typing.

  • Glass
Ordering information: Packaged in vials of 100 tubes.