Bunded spill flooring
Supplier: asecos
Certificates
About this item
The bunded spill flooring made from galavanised steel is designed for covering a large storage area.
- Removable, hot-dip galvanized grids
- Optimal adaption on space demands
- Approved for the safe storage of water pollutants and flammable liquids
Load capacity: 5 000 kg/m²
Frequently Bought Together