UV inspection lamp
Artikelnr. SIMELD7220
Supplier: Industrial Physics
Specifications
- Description:UV inspection spotlight, lamp head diameter 45 mm
- Intensity:1500 µW/cm²
- Pk:1
- Wavelength:365 nm
Specifications
About this item
This robust, handheld rechargeable inspection spotlight is used to detect contaminations that react under UV-illumination and cannot be seen with naked eye such as some organic fats and alkaline contaminants. Ideal to inspect the cleanliness of steel prior to painting.
- Equipped with high power UV LED (450 mW at 365 nm wavelength)
- Anodised aluminium lamp body is resistant to corrosion and stands up to years of heavy use
- Single wavelength, no heat but deep UV light
- Re-chargeable Li-ion battery with 180 min. life time
Output UV light intensity >1500 µW/cm² at 38 cm
Runtime (fully charged): approximately 3 hours
LED lifetime: 20000 hours
Weight: 190 g (without battery)
Delivery information: Supplied in a sturdy plastic suitcase complete with yellow safety glasses for optimal contrast, Li-ion battery, battery charger and carrying case.