Test kits, photometric, vicinal diketones, LCK cuvette tests
Catalog # HACHLCK242
Supplier: Hach
CAS Number:
Specifications
- Chemical method:MEBAK Method
- Pk:1 KIT
- Description:Vicinal diketones (VDK)
- Type:LCK242
- For unit:DR6000, DR5000, DR3900
- Range:0,015 - 0,5 mg/kg Diacetyl
Specifications
About this item
This ready-to-use, high-precision pre-dosed cuvette kit tests for the presence of vicinal diketones (VDK) with less hassle and no chemical waste. It also simplifies brewing analysis, with accurate results, every time.
- Provides high quality results free from potential external influencers
- Bar-coded cuvettes reduces errors and no reagent blank needed
- Innovative cuvette and reagent technique
- RFID technology to document the shelf life of the cuvettes and ensure traceability of your analyses
Storage conditions: 2 to 8 °C (keep refrigerated)