Luminescent bacteria test kit
About this item
The luminescent bacteria test is a simple operation to carry out as a chemical analysis. It provides quick results within maximum test time of 30 min.
- Long-life preserved bacteria
- Viability indicated by natural luminescence
- Effortless compensation for interferences
The preserved luminescent bacteria are reactivated before the test is started. Toxicity is indicated by the luminescence, more the sample inhibits luminescence, more toxic it is.
Certifications: Conforms to international EN ISO 11348 standard.
2 Options Available Below
Product Details & Documents
The luminescent bacteria test is a simple operation to carry out as a chemical analysis. It provides quick results within maximum test time of 30 min.
- Long-life preserved bacteria
- Viability indicated by natural luminescence
- Effortless compensation for interferences
The preserved luminescent bacteria are reactivated before the test is started. Toxicity is indicated by the luminescence, more the sample inhibits luminescence, more toxic it is.
Certifications: Conforms to international EN ISO 11348 standard.
Documents
Certificates
Search by Lot NumberHach LCK Cuvette Tests
Product Literature
HACHLCK482_TDS_EN
Datasheet
Product Family Options
Product Information
- PkAvailabilityPrice
- HACHLCK481NaCl solution 2% for luminescent bacteria test
- HACHLCK488TOX cuvette test with conserved luminescent bacteriaTOX cuvette test with conserved luminescent bacteria20 TestsLog in for availability1 * 20 Tests175,00 €