Orders LinkContactUsLinkComponent
Permeability cups
Permeability cups
Supplier:   Industrial Physics
Permeability cups
Supplier:  Industrial Physics
Certificates
Permeability cups
Permeability cups
2 Options Available Below
Product Details & Documents
Permeability cups are designed to determine the water vapour transmission of paints, varnishes, coatings and related products. Permeability cup is suitable for testing both self-supporting coating and non-self-supporting coatings.
  • Anodised aluminium for corrosion resistance and colouring
  • Seal ring to prevent turning when closing the cover
  • Any deformation to the cup may cause deviation in the result
The wet cup method is the reference for determining water vapour transmission.
Certifications: ISO 7783 (supersedes NF T30-018), ASTM D1653, ASTM E95.
Delivery information: Includes a cup, a seal ring and cover ring.
Documents
Product Family Options
Product Information
  • Availability
    Price
  • SIMEVF2200
    Permeability cup with a surface area of 10 cm²
    Permeability cup with a surface area of 10 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 235,00 €
    235,00 €
    1 * 1 ST
       235,00 €
  • SIMEVF2201
    Permeability cup with a surface area of 25 cm²
    Permeability cup with a surface area of 25 cm²
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 278,00 €
    278,00 €
    1 * 1 ST
       278,00 €