About this item
The Rapid Translation System (RTS) is a scalable in vitro protein expression platform that produces large amounts of protein for characterisation studies, functional assays, or structural analysis. RTS enables both transcription and translation utilising High-Yield (HY) and continuous-exchange, cell-free (CECF) technologies, with systems based on bacterial (E. coli) or eukaryotic (wheat germ) cell lysates. RTS offers the advantages of a cell-free protein expression system, whilst eliminating the need for laborious up and downstream steps typically associated with cell-based expression systems.
Wheat Germ Kits
RTS 100 Wheat Germ CECF Kit: Microgram-scale protein expression of functional eukaryotic proteins
Express eukaryotic proteins with high success
High yields due to microplate CECF device
Maximise the production of active heat-sensitive proteins, due to an optimum reaction temperature of 24 °C
Avoid cloning - express your protein directly from PCR products related to recombinant organisms
RTS 500 Wheat Germ CECF Kit: Milligram-scale protein expression of functional eukaryotic proteins
Cell-free expression of up to 1 mg functional eukaryotic protein
Express eukaryotic proteins with high success, generally without sequence optimisation
Maximise the production of active heat-sensitive proteins, due to an optimum reaction temperature of 24 °C