ContactUsLinkComponent
Precision scale magnifiers
Precision scale magnifiers
  ESCH155074
undefined
Precision scale magnifiers
  ESCH155074
  • Magnification:
    10×
  • Description:
    Magnifier with battery LED handle (Ø = 35 mm), 38,0 dioptre
  • Seq:
    03
  • Environmentally Preferable:
  • Pk:
    1
Precision magnifiers with brightness control and removable measuring scale available with battery or mains handle with brightness control.
Aspheric PXM® lightweight lens with Cera-tec® hard coating (both sides).