Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Xylene Cyanole FF
Xylene cyanole FF
Xylene cyanole FF
Xylene cyanole FF
Catalog # RC-115
Supplier:
G-Biosciences
CAS Number:
Expand image
Xylene cyanole FF
Catalog # RC-115
Supplier:
G-Biosciences
CAS Number:
Specifications
Pk:
10 g
Seq:
3236
Specifications
About this item
Product Details
:
:
538.62 g/mol
Melting Pt:
295 °C
CAS Number:
2650-17-1
Formula:
C₂₅H₂₇N₂NaO₆S₂
MDL Number:
MFCD00040657
Product Details