Order Entry
Austria
ContactUsLinkComponent
Magnetic thermometer
Magnetic thermometer
Catalog # SIMETM0015
Supplier:  Industrial Physics
undefined
Magnetic thermometer
Catalog # SIMETM0015
Supplier:  Industrial Physics

Specifications

  • Ø:
    65 mm
  • Description:
    Magnetic thermometer
  • Pk:
    1
  • Range:
    −10…+70 °C; +14...+60 °F

Specifications

About this item

Magnetic thermometer for measuring the surface temperature of steel objects. The thermometer can easily be placed on ferrous substrates with the strong magnet on the back.

  • Surface temperature can be read out in both °C and °F
  • Can be used in vacuum
  • No plastic parts on the outside of the thermometer
Delivery: Supplied in protective leather pouch with belt-clip.