Order Entry
Austria
ContactUsLinkComponent
Trisodium phosphate dodecahydrate ≥92%, Reagent Grade
Trisodium phosphate dodecahydrate ≥92%, Reagent Grade
Catalog # 0268-1KG
Supplier:  VWR Chemicals
CAS Number:  
undefined
Trisodium phosphate dodecahydrate ≥92%, Reagent Grade
Catalog # 0268-1KG
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Arsenic:
    ≤0.0005%
  • Heavy Metals:
    ≤0.005%
  • Insolubles:
    ≤0.2%
  • pH (10%, Water) @25 °C:
    ≥12
  • Purity (Dry Basis):
    ≥92%
  • Solubility (10%, Water):
    Pass

Specification Test Results

About this item

 :
 :380.12 g/mol
Melting Pt:∼75 °C
Density:1.62 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-509-8
UN:3268
ADR:8,III
 :GHS07
CAS Number:10101-89-0
Formula:Na₃PO₄·12H₂O
MDL Number:MFCD00149198