Order Entry
Austria
Orders LinkContactUsLinkComponent
Multi-parameter electrode (pH/ORP/conductivity), sensION+ 5048

Multi-parameter electrode (pH/ORP/conductivity), sensION+ 5048

Multi-parameter electrode, sensION™+ 5048, sensION™+ 5048 (pH/ORP/conductivity), 20 µS/cm - 200 mS/cm, 0 - 14, ±2000, 0...+80 °C, PC, Ceramic pin, Gel (non refillable), 12×85 mm, MP8, 1,5 m cable, General aqueous applications
Catalog # HACHLZW5048970002
Supplier:   Hach
Multi-parameter electrode (pH/ORP/conductivity), sensION+ 5048
Catalog # HACHLZW5048970002
Supplier:  Hach
Multi-parameter electrode (pH/ORP/conductivity), sensION+ 5048
Multi-parameter electrode (pH/ORP/conductivity), sensION+ 5048

Multi-parameter electrode, sensION™+ 5048, sensION™+ 5048 (pH/ORP/conductivity), 20 µS/cm - 200 mS/cm, 0 - 14, ±2000, 0...+80 °C, PC, Ceramic pin, Gel (non refillable), 12×85 mm, MP8, 1,5 m cable, General aqueous applications

Price
688,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Multisensor for pH, ORP, conductivity and temperature. For use with sensION+ MM150 multi-parameter meter.

  • Accurate
  • Easy to use
  • Rugged and durable
  • Automatic temperature compensation
  • Glass (pH); platinum (conductivity & ORP); and Pt1000 (temperature) sensors

Attachable calibration tubes can screw into handle of electrode (only 2 ml of buffer or standard). Storage protector prevents breakage and avoids over pressurisation of the diaphragm. Ideal for pH, conductivity, ORP and temperature measurements in general aqueous applications.

Specifications
  • Conductivity:
    20 µS/cm - 200 mS/cm
  • Connection:
    MP8, 1,5 m cable
  • Ø×L:
    12×85 mm
  • For:
    General aqueous applications
  • Type:
    sensION™+ 5048 (pH/ORP/conductivity)
  • Description:
    Multi-parameter electrode, sensION™+ 5048
  • Material:
    PC
  • Seq:
    02
  • mV:
    ±2000
  • Diaphr.:
    Ceramic pin
  • Electrolyte:
    Gel (non refillable)
  • Temp.:
    0...+80 °C
  • pH:
    0 - 14