ContactUsLinkComponent
Microscope slide with positive charge
Microscope slide with positive charge
Catalog # 631-1237
Supplier:  KLINIPATH
Microscope slide with positive charge
Catalog # 631-1237
Supplier:  KLINIPATH
Specifications
  • Description:
    Positively charged slide with frosted end and special barrier
  • Seq:
    02
  • Pk:
    72
Specifications
About this item
This microscope slide is designed with grounded-90, finished and clipped corners of 1 mm.
  • Suitable for use in all types of slide printers
  • Used for IHC staining and other special techniques
  • Easy handling in slide racks, automated slide printers and digital slide imagers
Packaging: In sealed plastic dispenser boxes.