Reference standard kits for use with MD 100 and MD 200 photometers
Supplier: Lovibond
Certificates
About this item
Reference standards are designed to check the accuracy and the reliability of the results.
- With a shelf life of two years
- Not possible to calibrate the photometer with the reference standards
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together