Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Sodium Chloride Standard Solution, 491 mg/L NaCl (1000 µS/cm)

Sodium chloride standard solution, 1000 µS/cm, 491 mg/l as NaCl, 1 L
Catalog # HACH1440053
Supplier:   Hach
Sodium Chloride Standard Solution, 491 mg/L NaCl (1000 µS/cm)
Catalog # HACH1440053
Supplier:  Hach
CAS Number:  
Sodium Chloride Standard Solution, 491 mg/L NaCl (1000 µS/cm)
Sodium Chloride Standard Solution, 491 mg/L NaCl (1000 µS/cm)

Sodium chloride standard solution, 1000 µS/cm, 491 mg/l as NaCl, 1 L

CAS Number:  
Price
102,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Standard solution for standardisation and accuracy check in conductivity measurements.

 

Specifications

Specifications

  • Konzentration:
    491 mg/l as NaCl
  • VE:
    1 L
  • Bezeichnung:
    Sodium chloride standard solution
  • Leitfähigkeit:
    1000 µS/cm

Specifications