Beryllium standard solution, for ICP-MS, Pure Plus grade
Artikelnr. PERCN9303730
Supplier: PerkinElmer
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:2% HNO₃
- Pk:125 mL
- Contents:1000 mg/l
- Element:Beryllium
- Symbol:Be
Specifications
About this item
Single element standard is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
- Analysed by ICP-OES
- Analysed by classical wet assay
- 67 trace impurities analysed by ICP-MS of the final solution and reported on the certificate
- Impurities reported at ppb level
- Prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Caution: These standards are in acid solution, for full details of hazards please read the accompanying Material Safety Data Sheets (MSDS).