Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Cationic Standards
Mercury Standards
Single element standards for ICP, mercury
Single element standards for ICP, mercury
Single element standards for ICP, mercury
Catalog # HEWL8500-6941
Supplier:
Agilent
CAS Number:
Expand image
Single element standards for ICP, mercury
Catalog # HEWL8500-6941
Supplier:
Agilent
CAS Number:
Specifications
Concentration:
10 mg/L
Matrix:
2% HNO₃
Pk:
1
Element:
Mercury
Symbol:
Hg
Specifications
About this item
Product Details
Product Details