Order Entry
Austria
ContactUsLinkComponent
Knee Protector, Standard
Knee Protector, Standard
Catalog # SIOEYMB00000C0
Supplier:  SIOEN
undefined
Knee Protector, Standard
Catalog # SIOEYMB00000C0
Supplier:  SIOEN

Specifications

  • Colour:
    Black
  • Description:
    Knee protector, standard
  • L×W:
    240×147 mm
  • Thickness:
    20 mm
  • Pk:
    1

Specifications

About this item

These knee protectors are recommended for work in the kneeling position. The protection distributes forces evenly and prevents small hard objects on the ground causing injuries.

Without knee protectors, workers may suffer discomfort and immediate injuries from hard surfaces and small stones and similar objects lying on the surfaces.

Certifications: Tested according to EN 14.404+A1:2010