Orders LinkContactUsLinkComponent
Immersion circulators, AC150
Immersion circulators, AC150
Supplier:   THERMO ELECTRON LED
Immersion circulators, AC150
Certificates
Immersion circulators, AC150
Immersion circulators, AC150
2 Options Available Below
Product Details & Documents
AC150 Immersion circulators are ideal for applications that require greater pumping performance with features such as ramp programming, application alarms and temperature ranges from −50 to 150 °C.
  • Automatic controller shutdown at detection of excessive high temperature, low liquid level or motor overload
  • Programmable application temperature alarm with user selected alarm, go-safe-state or shut off option
  • Five programmable set point temperatures and self-tuning PID controller for optimized temperature control
  • Low liquid level alarm for Safety Class III and RTA (Real Temperature Adjustment) for calibration
  • Powerful force and suction pump for external open and closed applications
Also features USB communication port and options for RS232, RS485, Ethernet/LAN, Analog I/O and internal or external temperature control mode (Remote Sensor, NAMUR type). Acoustic and optical alarm with auto restart feature after power failure.
Delivery information: It includes hose barbs with bridge (8 and 12 mm), pump plug for external circulation and power cord (1,8 m).
Product Family Options
Product Information
  • Type
    Temperature Range
    Temp. stability
    Heating capacity
    W×D×H
    Weight
    Availability
    Price
  • AC150
    13...200 °C
    0,01 °C
    2 kW/1,2 kW
    165×199×372 mm
    4,2 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 720,00 €
    2 720,00 €
    1 * 1 ST
       2 720,00 €
  • AC150 with bridge
    −50...+150 °C
    0,01 °C
    1,2 kW
    165×199×372 mm
    4,2 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 550,00 €
    2 550,00 €
    1 * 1 ST
       2 550,00 €