Order Entry
Austria
ContactUsLinkComponent
Immersion temperature probe
Immersion temperature probe
Catalog # GRSGGTF4011/3DIN
Supplier:  Greisinger
undefined
Immersion temperature probe
Catalog # GRSGGTF4011/3DIN
Supplier:  Greisinger

Specifications

  • Range:
    −50…+400 °C
  • Description:
    Immersion probe, 4-wire, 1/3 DIN class B
  • Seq:
    01
  • Pk:
    1
  • Ø×L:
    3×150 mm

Specifications

About this item

These measuring temperature probes are ideal for liquids and gases.

  • Stainless steel tube (V4A) with plastic handle
  • Non-corrosive
  • Anti-buckling glanding