VWR® Dropping funnels, cylindrical, without pressure balance
Supplier: VWR Collection
Certificates
About this item
Borosilicate 3.3 glass.
- Ground glass cone for flasks and assemblies
- PTFE key stopcock and PP stopper
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
PTFE key stopcock for non stick, grease-free performance.
Certifications: ISO 4800
Frequently Bought Together