Culture tubes, disposable, with screw thread finish, KIMBLE®
Supplier: DWK Life Sciences
Certificates
About this item
These culture tubes are manufactured from 51 expansion borosilicate glass. Suitable for tissue culture work and general bacteriological use.
- Round-bottomed
- Each tube has a chevron marking area
- Modular trays are shrink-wrapped in plastic film
- Supplied without caps
Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class B requirements.
Frequently Bought Together