ContactUsLinkComponent
Foam hand sanitiser dispenser, InstantFOAM TouchFREE
Foam hand sanitiser dispenser, InstantFOAM TouchFREE
Catalog # EVONIFSTF2MD-UK
Supplier:  Deb-STOKO
undefined
Foam hand sanitiser dispenser, InstantFOAM TouchFREE
Catalog # EVONIFSTF2MD-UK
Supplier:  Deb-STOKO
Specifications
  • Capacity:
    1000 ml
  • Colour:
    White
  • Description:
    Foam hand sanitiser dispenser, InstantFOAM TouchFREE, plastic
  • W×D×H:
    170×100×277 mm
  • Pk:
    1
Specifications
About this item
Energy-efficient touch-free dispensing system for use with foam hand cleansers.
  • Minimises any risk of cross-contamination
  • 4 D-cell batteries provide up to five years of operation
  • Infrared sensing technology to prevent misfiring
  • BioCote® protection antimicrobial agent inhibits the growth of bacteria and moulds
  • Translucent covers provide visibility of the cartridge without opening the dispenser
  • Dispenser can be locked and opened with the special key provided, can also be easily converted to a keyless, non locking operation
  • For wall-mounting
Suitable for use in public facility washroom, offices, medical rooms, laboratories, repair shops, food service and catering areas with InstantFOAM® cartridges.