Order Entry
Austria
ContactUsLinkComponent
Calibration standard, single element, tantalum for AAS and ICP-OES, pure grade
Calibration standard, single element, tantalum for AAS and ICP-OES, pure grade
Catalog # PERCN9303803
Supplier:  PerkinElmer
CAS Number:  
Calibration standard, single element, tantalum for AAS and ICP-OES, pure grade
Catalog # PERCN9303803
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O/0,8% HF
  • Pk:
    1
  • Element:
    Tantalum
  • Symbol:
    Ta

Specifications

About this item

Caution: These standards are in acid solution, for full details of hazards and specification, please see website and the links to Material Safety Data Sheets (MSDS) and Technical Data sheets.