Order Entry
Austria
ContactUsLinkComponent
 
Calibration standard, single element, neodymium for AAS and ICP-OES, pure grade
  PERCN9303787
 :  
Calibration standard, single element, neodymium for AAS and ICP-OES, pure grade
  PERCN9303787
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Pk Info:
    1× 125 ml
  • Element:
    Neodymium
  • Symbol:
    Nd

 

 

 : These standards are in acid solution, for full details of hazards and specification, please see website and the links to Material Safety Data Sheets (MSDS) and Technical Data sheets.