Order Entry
Austria
ContactUsLinkComponent
Calibration standard, single element, sodium for AAS and ICP-OES, pure grade
Calibration standard, single element, sodium for AAS and ICP-OES, pure grade
Catalog # PERCN9303785
Supplier:  PerkinElmer
CAS Number:  
Calibration standard, single element, sodium for AAS and ICP-OES, pure grade
Catalog # PERCN9303785
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Pk Info:
    1×125 ml
  • Element:
    Sodium
  • Symbol:
    Na

Specifications

About this item

Pure Grade standards for AA and ICP-OES.

  • Analyzed by ICP-OES
  • Analyzed by Classical Wet Assay
  • 32 trace impurities analyzed by ICP-MS of the final solution and reported on the certificate
  • Impurities reported at ppm level
  • All Standards are prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Caution: These standards are in acid solution, for full details of hazards and specification, please see website and the links to Material Safety Data Sheets (MSDS) and Technical Data sheets.