Order Entry
Austria
Orders LinkContactUsLinkComponent
Microplate reader, multi-mode, SpectraMax® Paradigm®

Microplate reader, multi-mode, SpectraMax® Paradigm®

SpectraMax® Paradigm®, microplate reader, multi-mode
Catalog # MLDVPARADIGM
Supplier:   Molecular Devices
Microplate reader, multi-mode, SpectraMax® Paradigm®
Catalog # MLDVPARADIGM
Supplier:  Molecular Devices
Microplate reader, multi-mode, SpectraMax® Paradigm®
Microplate reader, multi-mode, SpectraMax® Paradigm®

SpectraMax® Paradigm®, microplate reader, multi-mode

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

The SpectraMax® Paradigm® reader can perform absorbance, fluorescence, time-resolved fluorescence (including HTRF), fluorescence polarisation, AlphaScreen®, AlphaLISA®, and luminescence measurements. Detection cartridge modularity allows users to configure the system to meet current needs, while providing the flexibility to address future applications.

  • Flexible design allows for single or dual excitation and emission, to address existing needs and future expansion
  • User upgradable in <2 minutes
  • High speed detection
  • Excellent sensitivity and performance
  • SpectraDrop™ plate protocols enabling microvolume applications measurement are included
  • Patented LED intensity adjustment

Other features include an array of detection cartridge capabilities, automatic Z-height optimisation (both top- and bottom-read configurations), fluorescence measurements no longer require users to adjust the gain, up to 1536-well format, temperature control, monochromator-based absorbance, filter-based fluorescence, and TUNE Cartridge available for unmatched sensitivity in wavelength tunable detection.

Specifications
  • Description:
    SpectraMax® Paradigm®, microplate reader, multi-mode
  • Incubation range:
    4 °C above ambient to 45 °C
  • Light source:
    High powered light source
  • Temperature uniformity:
    ±0,75 °C
  • Photometric range:
    0,000 - 3,5 OD
  • Shaking:
    Linear, orbital
  • Wavelength bandwidth:
    4,0 nm
  • Photometric accuracy:
    0,010 OD ±2%
  • Wavelength accuracy:
    ±1 nm
  • Weight:
    45 kg
  • Cat. No.:
    MLDVPARADIGM
  • Plate format:
    6- to 1536-well
  • Wavelength selection:
    1,0 nm increments
  • Photometric linearity:
    0,0 - 2,0 OD
  • Model:
    SpectraMax® Paradigm®
  • Wavelength range:
    230 - 1000 nm
  • W×D×H:
    390×630×450 mm
  • Software:
    SoftMax® Pro
Complementary Products

Product Information

  • Catalog #
    Description
    Pk
    Serum format
    Sterility
    Packed
    Recommended working volume
    Culture area
    Sterile
    Colour
    Availability
    Price
  • 30026.
    Calcein-AM Cell Viability Assay kit, 1000 assays
    1 KIT
    Log in for availability
    1 * 1 KIT - 417,00 €

  • 30029.
    NucView™ 488 Caspase-3 Assay Kit for live cells, 100 assays
    1 KIT
    Log in for availability
    1 * 1 KIT - 631,00 €

  • S1810-500
    Foetal bovine serum, South American origin
    Frozen
    15
    Log in for availability
    1 * 500 mL - 330,00 €

  • 30002.
    Viability/Cytotoxicity Assay kit for Animal Live & Dead Cells, 300 assays
    1 KIT
    Log in for availability
    1 * 1 KIT - 440,00 €

  • 30009-2
    Caspase-3 DEVD-R110 Fluorometric HTS Assay Kit, 10 ml
    1 KIT
    Log in for availability
    1 * 1 KIT - 294,00 €

  • 31006.
    AccuBlue® High sensitivity dsDNA quantitation kit with 8 DNA standards
    1.000 Assays
    Log in for availability
    1 * 1.000 Assays - 435,00 €

  • 31007.
    AccuBlue® Broad range dsDNA quantitation kit with 9 DNA standards
    1.500 Assays
    Log in for availability
    1 * 1.500 Assays - 435,00 €

  • 31028.
    AccuClear® Ultra high sensitivity dsDNA quantitation kit with 7 DNA standards
    1.000 Assays
    Log in for availability
    1 * 1.000 Assays - 518,00 €

  • 734-2327
    1/bag
    0,075 - 0,20 ml
    0,33 cm²
    +
    17
    Clear
    Log in for availability
    1 * 100 ST - 243,00 €